Tlačiť
Storage and Stacking Containers
Dodávateľ: BURKLE
3414-0117 3414-0111 3414-0164 3414-0138 3414-0148 3414-0120 3414-0181 3414-0123
216-1723EA 34.89 EUR
216-1723 216-1724 BURK3414-0164 BURK3414-0138 BURK3414-0148 BURK3414-0120 BURK3414-0181 BURK3414-0123
Storage and Stacking Containers
Krabice Krabice přepravní
PP, grey.
  • Temperature resistant from −40 to +90 °C
  • According to Euro standards
  • Lid and transport rollers available as accessory
  • Suitable for use with foodstuffs
Stable storage containers with external reinforcement ribs.