Tlačiť
Tissue embedding cassettes, UNI type
Dodávateľ: KARTELL
2921-03 2921-04 2921-06 2921-12 2921-13 2921-10
631-9610EA 126.75 EUR
631-9610 631-9611 631-9613 631-9614 631-9615 KART2921-10
Tissue embedding cassettes, UNI type
Kazety pro zalévání vzorků Kazety tkáňové
Tissue embedding cassettes for paraffin inclusions, Universal type, provided with clip on closure lid that can easily be removed. They are available in five colours. Dimensions: 28×40×6,8 mm.