Tlačiť
BD™ Quincke Spinal Needles
Dodávateľ: BD Medical
405258 405248 405253 405259 405250 405257 405256 405247 405240 405252 405255
BDAM405258EA 105.5 EUR
BDAM405258 BDAM405248 BDAM405253 BDAM405259 BDAM405250 BDAM405257 BDAM405256 BDAM405247 BDAM405240 BDAM405252 BDAM405255
BD™ Quincke Spinal Needles
Jehly ke stříkačkám
BD® Quincke Spinal Needles are available in different sizes with lengths ranging from 90 to 178 mm and gauges from 18 to 27 G. The key/slot arrangement of stylet and cannula hubs on the BD® Quincke Spinal Needles facilitates proper needle bevel orientation.
  • Enhanced precision with BD® Quincke Spinal Needles that enable visualisation of cerebrospinal fluid (CSF), aiding proper needle placement
  • Fitted stylet designed to reduce tissue coring
  • Translucent window hub featuring contact clarity that helps allow visualisation of CSF
Balenie: Packaged 25 per box, with 8 boxes per case (total 200).