Tlačiť
Dropping funnels, cylindrical, with pressure balance, DURAN®
Dodávateľ: DWK LIFE SCIENCES
241252009 241252403 241252506 241253605 241253708 241254404 241254601
SCOT241252009EA 183.31 EUR
SCOT241252009 SCOT241252403 SCOT241252506 SCOT241253605 SCOT241253708 SCOT241254404 SCOT241254601
Dropping funnels, cylindrical, with pressure balance, DURAN®
Nálevky Nálevky přikapávací
DURAN®, borosilicate glass 3.3, clear, with glass key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket, with divisions for approximate volume reading.
* non-DIN ISO size
Certifikácie: DIN ISO 4800