Tlačiť
Syringes, BD PosiFlush™
Dodávateľ: BD Medical
306573 306574 306572
BDAM306573EA 562.78 EUR
BDAM306573 BDAM306574 BDAM306572
Syringes, BD PosiFlush™
Stříkačky Střílkačky zdravotnické
BD PosiFlush™ is a range of pre-filled saline syringes containing sodium chloride which are specifically designed for flushing in-situ vascular access devices in order to help maintain patency and reduce the risk of contamination.
  • Available in two options (BD PosiFlush™ SP and BD PosiFlush™ XS)
BD PosiFlush™ SP: Sterile Fluid Path only.
BD PosiFlush™ XS: Externally Sterile and can be used on a sterile field.