Tlačiť
Sampler, Tubus
Dodávateľ: BURKLE
5382-0080 5382-0040
223-1252EA 217.5 EUR
223-1252 223-0083
Sampler, Tubus
Vzorkovače Vzorkovače pevné
Sampler for free flowing bulk goods up to approx. 10 mm diameter. This makes it possible, for example, to remove hazel nuts, blends of tea, grain or similar items from large sacks.
  • Also for coarse grained bulk goods
  • Made of stainless steel V4A (1.4404)/AISI 316L
  • Special tip geometry
Corresponds to ISTA standard for seed lengths of 10 mm (wheat, oats) and 5 mm (flax, vetches).