Tlačiť
Culture tubes, disposable, with straight rim, DURAN®
Dodávateľ: DWK LIFE SCIENCES
231720965 231720998 231721197 231721283 231720184 231720784 231721883 231721986 231721489 231722193 231720365 231720862 231721094
SCOT231720965EA 37.02 EUR
SCOT231720965 SCOT231720998 SCOT231721197 SCOT231721283 SCOT231720184 SCOT231720784 SCOT231721883 SCOT231721986 SCOT231721489 SCOT231722193 SCOT231720365 SCOT231720862 SCOT231721094
Culture tubes, disposable, with straight rim, DURAN®
Zkumavky Zkumavky kultivační
Soda-lime glass with straight rim.
For use in applications without high thermal loads.