Tlačiť
Microcentrifuge tubes, 0,6/1,5/2,0 ml, graduated, with snap cap
Dodávateľ: Thermo Fisher Scientific
3448 3434 3449 3451 3453 3446
525-1502EA 20.58 EUR
525-1502 525-1503 525-1516 525-1517 525-1518 525-0468
Microcentrifuge tubes, 0,6/1,5/2,0 ml, graduated, with snap cap
Zkumavky Mikrozkumavky centrifugační
These snap cap tubes are made with ultra clear virgin PP offering excellent clarity allowing for simple sample retrieval.
  • Can withstand centrifugation to 30000×g (0,6 ml), 26000×g (1,5 ml) and 25000×g (2,0 ml)
  • Graduated, frosted
These tubes can be boiled, autoclaved at 120 °C at 15 psi for 15 minutes, and stored at −80 °C.
Certifikácie: Non sterile: Certified DNA, Rnase/Dnase, pyrogen and endotoxin-free
Sterile: Certified DNA, Rnase/Dnase, pyrogen, endotoxin, PCR inhibitor, and bioburden-free