Tlačiť
Separating funnels, spherical, DURAN®
Dodávateľ: DWK LIFE SCIENCES
242913608 242914407 242915403 242916605 242917301 242918606
527-1411EA 194.27 EUR
527-1411 527-1412 527-1413 527-1414 527-1415 SCOT242918606
Separating funnels, spherical, DURAN®
Nálevky Nálevky dělící
DURAN® borosilicate glass 3.3, clear, stopcock with glass key and plastic stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance