Tlačiť
Intravenous catheter, Surflo®
Dodávateľ: TERUMO
SR-DM1445PX SR-DM1645PX SR-DM1645WX SR-DM1745PX SR-DM1832PX SR-DM1832WX SR-DM1845PX SR-DM1845WX SR-DM2032PX SR-DM2032WX SR-DM2225PX SR-DM2225WX SR-DM2419PX SR-DM2419WX SR-DM2619PX SR-DM2619WX SR-OX1664C1 SR-OX1464C1 SR-OX1651C1 SR-OX1832C1 SR-OX1851C1 SR-OX1864C1 SR-OX2032C1 SR-OX2051C1 SR-OX2225C1 SR-OX2419C1
TERUSR-DM1445PXEA 175.29 EUR
TERUSR-DM1445PX TERUSR-DM1645PX TERUSR-DM1645WX TERUSR-DM1745PX TERUSR-DM1832PX TERUSR-DM1832WX TERUSR-DM1845PX TERUSR-DM1845WX TERUSR-DM2032PX TERUSR-DM2032WX TERUSR-DM2225PX TERUSR-DM2225WX TERUSR-DM2419PX TERUSR-DM2419WX TERUSR-DM2619PX TERUSR-DM2619WX TERUSR-OX1664C1 TERUSR-OX1464C1 TERUSR-OX1651C1 TERUSR-OX1832C1 TERUSR-OX1851C1 TERUSR-OX1864C1 TERUSR-OX2032C1 TERUSR-OX2051C1 TERUSR-OX2225C1 TERUSR-OX2419C1
Intravenous catheter, Surflo®
Kanyly
This IV catheter exists in three types: Straight (EFTE), wings (FEP), and wings and injection port (FEP).
  • Catheter tip is perfectly tapered to ensure a smooth transition from needle to catheter
  • Because the material is stronger than other materials, there is less likeliness of split tips and peelback
  • For almost each gauge an obturator is available with the same colour as the catheter
  • Obturator prevents thrombus formation in the lumen of the catheter when the infusion is discontinued