Tlačiť
pH electrodes for pharmaceutical and chemical industries, EasyFerm Plus
Dodávateľ: HAMILTON BONADUZ
238633-1553 238633-1213 238633-1243 238633-1253 238633-1273 238633-1223 238633-1263 238633-1151 238633-1171 238633-1141 238633-1111 238633-1523 238633-1161 238633-1573 238633-1533 238633-1513 238633-1543 238633-1121 238633-1112 238633-1142
662-2064EA 635.78 EUR
662-2064 662-2052 662-2053 662-2054 662-2055 662-2056 662-2057 662-2058 662-2060 662-2059 662-2061 662-2062 662-2063 662-2065 662-2066 662-2067 662-2068 662-2320 HAMI238633-1112 HAMI238633-1142
pH electrodes for pharmaceutical and chemical industries, EasyFerm Plus
Elektrody Elektrody pH
The EasyFerm Plus electrodes are designed to withstand demanding applications in pharmaceutical and chemical industries. They are supplied with a pre-pressurised electrolyte which prevents the diffusion of sample into the sensor.
  • Shaft material: Glass
  • pH range: 0 to 14
  • Pressure range: 0 to 6 bar
  • Electrolyte: Pressurised Phermlyte
  • Diaphragm: HP-Coatramic
The Everef-F reference cartridge ensures that the Phermlyte reference electrolyte remains free of silver and precipitation. The electrodes withstand steam sterilisation, autoclavation and CIP cleanings.
Certifikácie: ATEX / IECEx approval: CE 0035 ll 1/2 G Ex ia llC T4/T5/T6 Ga/Gb, CE 0035 ll 1/2 D Ex ia lllC T x °C Da/Db.