Tlačiť
Stainless steel fittings, Upchurch Scientific®
Dodávateľ: UPCHURCH SCIENTIFIC
U-320X U-321X U-400X U-401 U-401X U-410 U-410X U-450
554-1806EA 143.6 EUR
554-1806 554-1807 554-1808 UPCHU-401 554-1809 554-5694 554-1810 554-5695
Stainless steel fittings, Upchurch Scientific®
Fitinky a konektory
Note: Do not use metal fittings in plastic ports, as this can damage the port.
  • For coned ports
  • Rated to 20000 psi (1380 bar)