Tlačiť
Connector, glass, cone on socket, with stopcock
Dodávateľ: LENZ LABORGLAS
05310011 05310044
LENZ05310011EA 93.06 EUR
LENZ05310011 LENZ05310044
Connector, glass, cone on socket, with stopcock
Komponenty pro destilaci Adaptéry vícenásobné
DURAN® Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Connectors with NS stopcock, with solid glass plug, with NS socket and NS cone.