Tlačiť
Chromatographic columns with frit, cone and socket
Dodávateľ: LENZ LABORGLAS
05450901 05450902 05450912 05450914 05450916 05450904 05450906 05450911
LENZ05450901EA 153.63 EUR
LENZ05450901 LENZ05450902 LENZ05450912 LENZ05450914 LENZ05450916 LENZ05450904 LENZ05450906 LENZ05450911
Chromatographic columns with frit, cone and socket
Kolony chromatografické
Chromatography columns, DURAN® glass, with NS socket and NS cone.
  • With sintered frit - porosity zero
  • Suction tube thread GL 18