Tlačiť
Funnels with short stem
Dodávateľ: LENZ LABORGLAS
05318003 05318012 05318033 05318005 05318008 05318009 05318015 05318017 05318020 05318025
LENZ05318003EA 3.57 EUR
LENZ05318003 LENZ05318012 LENZ05318033 LENZ05318005 LENZ05318008 LENZ05318009 LENZ05318015 LENZ05318017 LENZ05318020 LENZ05318025
Funnels with short stem
Nálevky Nálevky filtrační Nálevky k filtraci
Soda-lime glass.
  • Excellent chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With smooth internal walls. Flare angle 60°.