jodmetan ≥99.5% stabilised
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
Synonymá:
Methyl iodide
Stabilizácia:
Stabilised with copper
Stabilized with copper
Purity: 99.5%
CAS No.: 74-88-4
Molecular Formula: CH3I
Formula Weight: 141.94
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 2.28
Refractive Index: 1.5300
Sensitivity: Light Sensitive
Appearance: Colourless to pale yellow
Odour: Pungent
Melting Point: -67°
Boiling Point: 41-43°
Beilstein Reference: 969135
Merck Reference: 14,6087
UN No.: 2644
MDL No.: MFCD00001073
Vzorec:
CH₃I Teplota varu: 41…43 °C Teplota tavenia: –67 °C Hustota: 2,28 Skladovacia teplota: Refrigerator |
Číslo MDL:
MFCD00001073 Číslo CAS: 74-88-4 EINECS: 200-819-5 OSN: 2644 ADR: 6.1,I Merck Index: 14,6087 |