Tlačiť

Heptafluorobutyryl chloride, Sigma-Aldrich®

Dodávateľ: Merck
Nebezpečí

Synonymá: Perfluorobutyryl chloride, 2,2,3,3,4,4,4-Heptafluorobutyryl chloride

M-Clarity™ quality level = MQ100

257923-25G 257923-5G
SIAL257923-25GEA 130.77 EUR
SIAL257923-25G SIAL257923-5G
Heptafluorobutyryl chloride, Sigma-Aldrich®
Heptafluorobutyryl chloride
Vzorec: C₄ClF₇O
Teplota varu: 39 °C (1013 hPa)
Hustota: 1,55 g/cm³ (25 °C)
Číslo MDL: MFCD00000707
Číslo CAS: 375-16-6
EINECS: 206-785-8
OSN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR