Tlačiť
Pipette cleaning sets, Nalgene®
Dodávateľ: Thermo Fisher Scientific
5250-0040 5250-0050 DS5250-0060
NALG5250-0040EA 1186.12 EUR
NALG5250-0040 NALG5250-0050 NALGDS5250-0060
Pipette cleaning sets, Nalgene®
Pipety Promývačky pipet
These conveniently packaged sets contain a basket, a jar and a washer. A larger size is available to accomodate up to 32" (81 cm) long pipettes.
  • All parts are made of rugged, high-density PE
  • Components are also available separately except F size (sold only as a set)