Tlačiť
Magnetic stirring bars, PTFE, cylindrical, with centre ring
Dodávateľ: BRAND
137404 137405 137410 137415 137420 137425 137427 137430 137432 137435 137445
BRND137404EA 75.01 EUR
BRND137404 BRND137405 BRND137410 BRND137415 BRND137420 442-9321 BRND137427 BRND137430 442-9855 BRND137435 BRND137445
Magnetic stirring bars, PTFE, cylindrical, with centre ring
Míchadla Míchadla magnetická
Cylindrical PTFE magnetic stirring bars with pivot ring.
  • Ideal for use with vessels that have slightly uneven bases
  • Pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position