Tlačiť
Microplate storage racks, Nunc™
Dodávateľ: Thermo Fisher Scientific
367002 344260 344280 344380
732-2756EA 271.56 EUR
732-2756 NUNC344260 NUNC344280 732-1571
Microplate storage racks, Nunc™
Stojany Stojánky pro mikrodestičky
A range of storage racks for standard microplates and DeepWell™ plates in chest or upright mechanical freezers or incubators.
Plastic racks
Freezer compatible down to −80 °C
Lightweight durable propylene and metal construction
Locking bar keeps plates secure and handle allows convenient transport
Aluminium racks
Adjustable shelves to fit any plate or tube rack
Autoclavable
Colour coded top and sides for easy identification
Lightweight with handle for ease of transport
Locking bar keeps plates secure