Tlačiť
Scoops, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
F367530000 F367560000 F367480000 F367490000 F367470000 F367500000
231-0397EA 71.4 EUR
231-0397 231-0398 BELAF367480000 231-2419 BELAF367470000 BELAF367500000
Scoops, SP Bel-Art
Lopatky
PP, translucent. These plastic scoops are rigid, strong and smooth. A flat bottom keeps them upright when filled or empty, allowing them to be used as weighing boats or to be placed on a flat surface.
  • 60 and 125 ml scoops have a longer handle that increases the distance between the hand and sample
  • Rugged PP; steam autoclavable at 121 °C
  • Comply with FDA regulations for food contact
The sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.