Tlačiť
Wireless multi-channel temperature monitoring systems, EBI 40
Dodávateľ: EBRO
1340-6400 1340-6401
620-1802EA 1799.44 EUR
620-1802 620-1803
Wireless multi-channel temperature monitoring systems, EBI 40
Systémy monitorování Systémy monitorování CO2 a teploty
EBI 40 loggers are designed for mapping applications at high temperatures. In combination with thermal insulation box (620-1804), they can be used for 2 hours at 250 °C. At higher temperatures, the logger can be placed remotely and temperature characteristics viewed on the display.
  • K-type thermocouple with SMP connector
  • Alternative programming via keypad buttons: ESC, OK, Up, Down
  • Configuration, program upload and data download via USB connection
  • Coloured display for numerical and graphical presentation of data
This system can be used in heating chambers and ovens, for determination of hardening (lacquering), and monitoring of complete baking and freezing processes in the food industry.
W×D×H: 140×118×35 mm.
Conforms to IP 67
Certifikácie: Conforms to IP 40.
Informace o dodávce: Supplied with a factory calibration certificate. Probes are not included.