Tlačiť
Cotton jersey gloves
Dodávateľ: UNIGLOVES AERTZE UND KLINIKBED
13111 13100 13112 13113 1360 560-11 1380 1390
112-1728EA 40.91 EUR
112-1728 112-1727 112-1729 112-1730 112-1723 112-1724 112-1725 112-1726
Cotton jersey gloves
Tricot gloves made of 100% cotton jersey.
  • Bleached white
  • Hand-specific for great fit
Length: 240 mm
Colour: White
Cat. I
Certifikácie: Cat. I