Tlačiť
Fibronectin culture dishes, Corning® BioCoat™
Dodávateľ: Corning
354403 354451
734-0111EA 266.37 EUR
734-0111 734-0137
Fibronectin culture dishes, Corning® BioCoat™
Misky kultivační Misky pro tkáňové kultury
PS coated with a uniform application of human fibronectin. Promotes cell attachment, spreading, proliferation and diffentiation of many cell types, particularly fibroblasts and other mesenchymally-derived cells.
Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí ISO 14001 - Bedford