Tlačiť
Accessories for OLS200 and GLS Aqua Plus shaking water baths
Dodávateľ: GRANT INSTRUMENTS
SBT14 SBT28
462-8350EA 135.39 EUR
462-8350 462-8351
Accessories for OLS200 and GLS Aqua Plus shaking water baths
Lázně Lázně vodní třepací
A range of different stainless steel accessories for use with OLS and GLS Aqua Plus shaking water baths.
  • Universal flask tray secures various size containers from 25 ml to 1000 ml
  • Tray for test tube racks, holds up to five test tube racks
  • Test tube racks in various formats, these are perforated to aid convection
  • Perforated tray enables shaking baths to be used as unstirred baths
  • Sloping lids direct condensation down the side of the bath, protecting vessels in the bath. PC lids are supplied as standard with GLS Aqua Plus and OLS200 models
Pozor: * TestMore Product Information