Tlačiť
Vacuum aspiration systems, VACUBOY
Dodávateľ: INTEGRA BIOSCIENCE
155500 155510
612-0683EA 307.26 EUR
612-0683 612-0684
Vacuum aspiration systems, VACUBOY
Čerpadla Vakuové aspirační systémy
The VACUBOY hand operator is ideal for aspirating and suctioning liquids from cuvettes, microtitration plates, test tubes, Petri dishes, cell culture bottles, roller bottles, bioreactors and other typical laboratory vessels in a safe and controlled way. It is designed to be used with many different stainless steel aspiration adapters, Pasteur and serological pipettes and pipette tips for an enormous range of applications. Each adapter has been specially designed to match the dimensions and the geometry of the corresponding glass or plastic laboratory vessels. The hand operator can be connected to any vacuum source already in the laboratory, thus serving to improve the existing facilities.
  • Extremely easy, time-saving operation
  • Ergonomic design takes the strain out of tedious, repetitive aspiration tasks
  • All parts that come into contact with liquid are autoclavable
  • Can be attached to any vacuum source (e.g. water jet pumps, external vacuum pumps, house vacuum system)
  • A wide selection of single-channel and multi-channel adapters are available as options
VACUBOY hand operator:
The basic device with the technical features described above, a rubber adapter for Pasteur pipettes and a 40 mm long, stainless steel single-channel adapter.
VACUBOY hand operator set:
Includes the basic device as described above, a matching stand, rubber adapter for Pasteur pipettes, one single-channel and one 8-channel adapter, both stainless steel and 40 mm long, one single-channel plastic adapter for tips with or without an ejection mechanism, and one 8-channel plastic adapter for tips with an ejection mechanism.
Includes the basic device as described above, a matching stand, rubber adapter for Pasteur pipettes, one single-channel and one 8-channel adapter, both stainless steel and 40 mm long, one single-channel plastic adapter for tips with or without an ejection mechanism, and one 8-channel plastic adapter for tips with an ejection mechanism.