Tlačiť
Internal centering rings for small flange connections
Dodávateľ: VACUUBRAND
20660120 20660121 20660122 20660124 20660125 20660126
185-1221EA 20.06 EUR
185-1221 185-1223 185-1225 185-1222 185-1224 185-1226
Internal centering rings for small flange connections
Těsnění Těsnící kroužky
Stainless steel, with FPM O-ring seal.
  • Suitable for use in a temperature range from −40 to +150 °C
  • Can also be used for bakeable flanges in high vacuum engineering
  • FPM O-ring seal with low gas discharge