Tlačiť
Small flanges with hose nozzle
Dodávateľ: VACUUBRAND
20662500 20662510 20662511 20662516 20662517 20662518 20662519 20662521 20662522 20662523 20662806 20662807 20662808
185-2251EA 27.92 EUR
185-2251 185-2252 189-0032 185-2253 185-2254 181-0490 185-2256 185-2257 185-2258 185-2259 181-0154 181-0491 185-0000
Small flanges with hose nozzle
Fitinky a konektory
These flanges are made from aluminium or plastic.
  • Fast, easy assembly and disassembly to/from pumps
  • Safe, vacuum tight connection with optional centring and clamping rings
  • A wide range of flanges with hose nozzles for various internal hose diameters are available
For connecting pumps to vacuum hoses.