Tlačiť
Food thermometer, HACCP, testo 106
Dodávateľ: TESTO
0563.1063 0560.1063
620-2358EA 70.6 EUR
620-2358 620-3055
Food thermometer, HACCP, testo 106
Teploměry Teploměry digitální
This thermometer provides a fine, yet robust, measuring tip that is ideally suited for fast temperature monitoring and food inspection in hotels, large kitchens and supermarkets. The special food probe makes an almost invisible penetration hole.
  • Threshold manually set by user
  • Audible and optical alarm when limits are exceeded
  • Automatic recognition of final value: 'HOLD' function
Probe (Lר: 55×3 mm)
Informace o dodávce: Supplied with probe protecting cover and three-volt button cell battery.