Tlačiť
Overhead stirrer tools, propeller-type impellers
Dodávateľ: Heidolph Instruments GmbH & Co.KG
509-30000-00 509-31000-00 509-32000-00 509-33000-00 509-39000-10
441-1229EA 140.39 EUR
441-1229 441-1230 441-1231 441-1232 HEID509-39000-10
Overhead stirrer tools, propeller-type impellers
Míchadla Nástroje na míchání
Made from stainless steel.
  • For suspensions and homogenisation
  • Ideal for stirring liquids with medium to high viscosity
  • Create axial flow