Tlačiť
Overhead stirrer tools, propeller, centrifugal and paddle-type impellers
Dodávateľ: Heidolph Instruments GmbH & Co.KG
509-10000-00 509-11000-00 509-12000-00 509-13000-00 509-14000-00 509-18000-10
441-1223EA 86.24 EUR
441-1223 441-1224 441-1222 441-1225 441-1226 HEID509-18000-10
Overhead stirrer tools, propeller, centrifugal and paddle-type impellers
Míchadla Nástroje na míchání
Made from stainless steel.
  • For medium speed range
  • Ideal for stirring liquids with low to medium viscosity