Tlačiť
Mixing system, VISCO JET® for overhead stirrers
Dodávateľ: Heidolph Instruments GmbH & Co.KG
509-16121-00 509-16120-00 509-16081-00 509-16080-00 509-16060-00
441-0016EA 127.96 EUR
441-0016 441-0015 441-0014 441-0013 441-0012
Mixing system, VISCO JET® for overhead stirrers
Míchadla Nástroje na míchání
VISCO JET® homogenises low and high viscosity liquids in vessels by creating dynamic flow conditions by means of the so-called cone principle. Turbulent flows are created at the taper end by acceleration, displacement and retardation. These flows advance through the stirred medium and result in the new dynamic mixing motion.
  • Only one system for nearly every mixing task
  • No air intake, foaming or heating
  • Careful and gentle mixing
  • Short mixing periods
  • Less power consumption through low mixing speed
Informace o dodávce: Supplied with shaft.