Tlačiť
Suction samplers, Mini ViscoSampler
Dodávateľ: BURKLE
5341-1060 5341-1100 5341-1150
300-1032EA 382.67 EUR
300-1032 300-1027 300-1028
Suction samplers, Mini ViscoSampler
Vzorkovače Vzorkovače kapalné
Suction samplers for viscous media such as paste, sludge, cream, or oil.
  • Easy to clean
  • Sterilisable
  • Stainless steel V4A (1.4404)/AISI 316L
Designed for the pharmaceutical and cosmetics industries.
Informace o dodávce: Supplied with a 2 mm suction nozzle, which can be replaced with drills that have a wider aperture for sampling substances with higher viscosity.