Tlačiť
Laboratory mixers
Dodávateľ: WARING BLENDOR
8010EB 8010EG 8010ES 8011EB 8011EG 8011ES LB20EG LB20ES 8011BU
431-9332EA 1083.03 EUR
431-9332 431-9112 431-9113 431-9333 431-9110 431-9111 431-0106 431-0107 WARI8011BU
Laboratory mixers
Homogenizátory Míchačky laboratorní pro standardní analýzu
Reliable devices with powerful motors. For thorough mixing, dispersion and emulsification. Special cutter heads and high speed operation make these laboratory mixers suitable for many applications. Two basic devices with capacities of 1 or 2 L and a variety of stainless steel, heat resistant glass or PC beakers are available.
Informace o dodávce: Supplied with motor, cutter head, beaker and lid.