Tlačiť
Plastic folding magnifiers
Dodávateľ: ESCHENBACH OPTIK
11869 118210
630-8260EA 175.85 EUR
630-8260 630-8261
Plastic folding magnifiers
Skla zvětšovací /Lupy
These magnifiers featuring a modern design and a glass fibre reinforced PA body provide a distortion-free image.
  • For handicraft and technical applications
  • Dustproof body when closed
Ø: 23 mm