Tlačiť
UV viewing cabinets, Mini Chromato-Vue®
Dodávateľ: UVP ULTRA VIOLET PRODUCTS
95-0072-07 95-0072-01
630-0715EA 742.64 EUR
630-0715 630-0714
UV viewing cabinets, Mini Chromato-Vue®
Osvětlení/Lampy Komory pro UV zobrazení
The C-10 cabinet is designed for use with compact UV lamps (4 W, such as models UVGL-15 or UVGL-25, or handheld UV lamps (6 W), such as the UVGL-58. The C-15G Mini UV cabinet combines the C-10 cabinet with the handheld UVGL-58 multi-band UV lamp (254/365 nm).
  • Filter absorbs UV energy and protects eyes from harmful shortwave radiation
  • Comfortable soft rubber viewport with adequate space for eyeglasses
  • Lightweight access curtain provides easy entry and blocks out external light from cabinet interior
  • Removable bottom panel allows cabinet to be placed over large or bulky items for fluorescence inspection
Cabinet interior (L×W×H): 229×254×178 mm
Cabinet exterior (L×W×H): 229×267×305 mm