Tlačiť
Varné plotýnky
Dodávateľ: STUART SCIENTIFIC
04807-80 04807-70 04807-66
STUA04807-80EA 2758.92 EUR
STUA04807-80 STUA04807-70 STUA04807-66
Varné plotýnky
Ceramic top plate or aluminium/silicon alloy top plate.
  • 'Hot' warning light flashes whenever the plate temperature is above 50 °C
Ceramic: excellent chemical resistance, easy to clean and allows higher temperatures.
Aluminium/silicon alloy: efficient and fast rise in temperature.