Tlačiť
Shelving systems, Quick, MetroMax Q
Dodávateľ: METRO
MQ74PE/MQ1836G MQ74PE/MQ1848G MQ74PE/MQ1860G MQ74PE/MQ1872G MQ74PE/MQ2448G MQ74PE/MQ2460G MQ74PE/MQ2472G
132-3909EA 956.94 EUR
132-3909 132-3910 132-3911 132-3912 132-3914 132-3915 132-3916
Shelving systems, Quick, MetroMax Q
Nábytek kancelářský Police
Simple to configure using a clip lock system - no tools required.
  • The shelving height can be changed in just a few minutes
  • Supplied with four shelves as standard
  • Load capacity: From approx. 270 to 360 kg, depending on the size of the shelving system
Plastic coated steel and corrosion proof.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable