Tlačiť
Shelving systems, Metromax
Dodávateľ: METRO
MX74P/MX1848G MX74P/MX1860G MX74P/MX2460G
132-3902EA 1607.45 EUR
132-3902 132-3903 132-3907
Shelving systems, Metromax
Nábytek kancelářský Police
Tough polymer construction, edges manufactured out of stainless steel.
  • With four shelves each individually configured and easy to clean
  • Load capacity: From approximately 270 kg to 360 kg, depending on the size of the shelving system