Tlačiť
Elektroporační kyvety Plus
Dodávateľ: BTX
45-0124 45-0125 45-0126 45-0134
732-0020EA 306.06 EUR
732-0020 732-0021 732-0023 BTXI45-0134
Elektroporační kyvety Plus
Kyvety Kyvety elektroporační
High quality disposable cuvettes for transfection available in three sizes and compatible with electroporation systems.
  • Transfer pipette for manipulation of samples and gamma irradiated together for sterility
  • Polished aluminium electrodes in the cuvettes lowers arcing frequency
  • Cuvettes (without cap) are autoclavable
Cuvette caps are round for easy, one-handed removal, and are colour-coded for quick identification.