Tlačiť
Recirculating coolers, FC series
Dodávateľ: JULABO GMBH
9600063 9600123 9600126 9600160 9600060 9600120 9600163 9600166 9601256 9601060 9601063
473-3221EA 9107.1 EUR
473-3221 473-3231 473-3232 473-3240 473-3220 473-3230 473-3241 473-3242 473-0024 473-0038 473-0039
Recirculating coolers, FC series
Termostaty oběhové Termostaty oběhové Termostat cirkulační s chlazením
Environmentally friendly recirculating coolers that save tap water, units are rapid and powerful, and offer short cool-down times and high efficiency. Due to the compact design, the coolers fit either on or under laboratory benchtops. FC units feature two separate multi-displays (LED) for actual and set point values.
  • Built-in heating element to allow cooling and heating applications with FC models
  • FC coolers have an adjustable ratio for inlet/outlet temperature
  • Connection for external PT100 sensor for external measurement and control (T-type models)
  • RS232 interface, connections for standby input and alarm output
  • Connection for external programming, temperature recorder, external PT100 sensor (T-type models)
Models with the designation W are water cooled, operate almost noiselessly and offer reduced heat generation to the environment. The units feature permanent performance supervision via sensors and pump motor and compressor overload protection. In the event of any disturbance, units shut down completely. FC models have early warning systems with audible signal for low liquid level, and both units feature high/low temperature limits as well as freezing/dry running protection. Units are conveniently filled from the front and have a removable venting grid.
Max. flow rate / max. pressure:
FC models: 20 L/min, 500 mbar pressure
FC S models: 22 L/min, 1200 mbar pressure
FC T models: 28 L/min, 3500 mbar pressure
'W' denotes models are water cooled.
FC models: 20 l/min, 500 mbar pressure
FC S models: 22 l/min, 1200 mbar pressure
FC T models: 28 l/min, 3500 mbar pressure
Informace o dodávce: Supplied with 2 barbed fittings for 8 and 12 mm internal bore tubing. All units with an RS232 interface can use the free EasyTemp software for control via a PC; the software can be downloaded free of charge from the Julabo website.
Pozor: Note: Cooling capacity at  +20; +10; +5, −10 and −20 °C
FCW2500T with water cooling