Tlačiť
High temperature digital block heaters, BT5D
Dodávateľ: GRANT INSTRUMENTS
BT5D-16 BT5D-26
460-0063EA 5878.06 EUR
460-0063 460-0064
High temperature digital block heaters, BT5D
Termostaty blokové Termostaty blokové/ohřívací
High temperature digital block heaters with single fixed aluminium block. Units feature microprocessor control, digital temperature settings and display, high and low alarms, and a controlled heating rate. Heaters have LED with 0,1 °C setting resolution and 1 °C display resolution.
  • Operating temperature range is ambient +10 to 400 °C
  • Timer for operation from 1 to 9999 min, or continuous operation
  • Ideal for use at high temperatures - avoids the use of oil and laboratory remains steam-free
  • Temperature stability at 300 °C is ±0,5 °C and temperature uniformity is ±1% (maximum temperature difference between tubes: 2 at 200 °C, 3 at  300 °C, 4 at 400 °C)
  • Adjustable overtemperature cut-out
Pozor: * RT = Ambient
Note: Temp. stability value shown above is at 300 °C.