Tlačiť
Interchangeable blocks for block heaters, QBA, QBD and QBH
Dodávateľ: GRANT INSTRUMENTS
QB-13 QB-50 QB-0 QB-10 QB-12 QB-16 QB-18 QB-24 QB-E0 QB-E1 QB-E2 QB-H QDP-FL QDP-H QB-17H
460-8211EA 194.27 EUR
460-8211 460-8212 460-8510 460-8511 460-8512 460-8513 460-8514 460-8515 460-8516 460-8517 460-8123 460-8124 460-0084 460-0085 460-0182
Interchangeable blocks for block heaters, QBA, QBD and QBH
Termostaty blokové Bloky
Grant blocks have a depth of 63 mm, ensuring precise temperature control for test tube applications.
  • Suitable for 0,2 ml tubes, strips or microplates
  • Can be used with standard/high temperature 96-well microtitre plates
Single block W×D×H (mm): 140×50×63
Double-size interchangeable blocks for QBA2, QBD2 and QBH2 only.
Particularly useful in molecular biology and biotechnology fields.
QDP-H blocks (460-0085) for 0,2 ml microplates, strips or individual tubes have 96 holes in microplate configuration. The block accommodates one 96-well plate, 8×12-well or 12×8-well strips or up to 96 individual capped tubes. A snug fit is ensured in the precision drilled wells. Uniformity is ±0,3 °C within tubes across the block.
QDP-FL universal block (460-0084) for standard 96-well plate accommodates the most commonly used types of standard or high temperature plates and cell culture plates. Supplied with hinged lid. Uniformity is ±0,5 °C between the wells.
Informace o dodávce: Supplied with block extraction tool.
Pozor: These alternative double-size blocks are available for use with models QBD2, QBD4, QBH2 and QBA2 only.