Tlačiť
Printer and balance accessories
Dodávateľ: OHAUS
80850000 80850043 21253677 00410024 12101511 80252038 80251993 80252580 80252581 80252584 80251931 80251930 80500552 83021082 83021083 83021085 83021102 21203204 30529322 12120799 30059316 30063920 30064202 30093334 12120761 30628012 30467918 30529323
611-5111EA 86.92 EUR
611-5111 611-0598 611-1525 611-2423 611-4299 611-3887 611-5412 611-2540 611-2541 611-2543 611-2538 611-2537 611-2574 611-2416 611-2417 611-2419 611-2421 611-3748 611-2579 611-2580 611-2572 611-2622 611-2573 611-3751 OHAU12120761 611-5061 OHAU30467918 OHAU30529323
Printer and balance accessories
Váhy
The SF40A impact printer and the STP103 thermal printer are compatible with any Ohaus balance with an RS232 interface. They are particularly useful when coupled to an Ohaus balance with GLP functionality as they can provide a hard copy audit trail. A printer cable is required (but not supplied) for specific Ohaus balances.
Objednávacie informácie: There are a wide variety of accessories and replacement parts for Ohaus balances and the most commonly used are listed below: For further items please visit the local Avantor website.