Tlačiť
pH electrodes, InLab®, Expert Series
Dodávateľ: METTLER TOLEDO
51340288 51343100 51343101 51343102 51343103 51343104 51343105
662-2738EA 473.33 EUR
662-2738 662-2839 662-2840 662-2841 662-2842 662-2843 662-2844
pH electrodes, InLab®, Expert Series
Elektrody Elektrody pH
Ideal for critical samples such as suspensions or samples of unknown composition (e.g. wastewater).
  • ARGENTHAL™ reference system with silver-ion trap
  • Solid XEROLYT® polymer electrolyte: No need for a junction (no possibility of contamination)
  • Unbreakable PEEK shaft
  • Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
pH range: 0 to 14
Temperature range: 0 to 100 °C
Temperature probe: NTC30 kOhm for InLab® Expert Pro and InLab® Expert NTC30; PT1000 for InLab® Expert DIN and InLab® Expert PT1000
Objednávacie informácie: Supplied with test certificate.