Tlačiť
pH electrodes, SenTix®
Dodávateľ: WTW
103630 103631 103632 103633 103635 103636 103639 103640 103641 103642 103644 103645 103646 103647 103648 103650 103637 103643 103695 103651 103652 103660 103661 103664 103731 103732 103733 103744 103792 103805 103806 103807 103808 103809 103810 103811 103812 103813 103814 103815 103816 103817
662-1301EA 158.84 EUR
662-1301 662-1302 662-1303 662-1304 662-1305 662-1306 662-1308 662-2151 662-1309 662-1315 662-1317 662-0291 662-1318 662-1319 662-1400 662-1213 662-1307 662-1316 662-1277 662-0106 662-1871 662-1275 662-1872 662-2266 662-1682 662-1999 WTWA103733 WTWA103744 WTWA103792 WTWA103805 WTWA103806 WTWA103807 WTWA103808 WTWA103809 WTWA103810 WTWA103811 WTWA103812 WTWA103813 WTWA103814 WTWA103815 WTWA103816 WTWA103817
pH electrodes, SenTix®
Elektrody Elektrody pH
These proven and tested pH electrodes are easy to use and offer high quality and precision.
  • Electrodes with gel electrolyte and a plastic body are robust and shock-proof, the membrane glass is safeguarded by a protective shield
  • Electrodes with liquid electrolyte: Capillary-effect of the platinum wire guarantees constant outflow and prevents contamination by precipitates or even dirt
  • SenTix® HW and HWS electrodes: Electrolyte flow speed can be adjusted for optimal use
Optimised membrane glass guarantees quick measurement even at low temperatures. The easy to use fill-hole cover prevents electrolyte from leaking out during storage.
* with integrated temperature probe (NTC 30 KOhm).
** without cable (available separately).