Tlačiť
Dispensers for towel rolls, ZetMatic®
Dodávateľ: ZVG BADEN WUERTEMBERG
60965 60972-12
115-2790EA 107.5 EUR
115-2790 ZVGA60972-12
Dispensers for towel rolls, ZetMatic®
Zásobníky na ubrousky
For towel rolls (i.e. Cat. No. 115-2791 to 115-2796) or folded towels (i.e. Cat. No. 115-2750 to 115-2757).
  • Durable plastic
  • Easy installation
  • Lockable
Colour: White