Tlačiť
Dispensers for wipes and cloths, Tork W1 / W2 / W3 / W4
Dodávateľ: Essity
207210 652000 652008 652100 652108 653000 653008 654000 654008
115-0335EA 101.48 EUR
115-0335 115-1265 115-1399 115-1266 115-1400 115-1267 115-1268 115-1269 115-1401
Dispensers for wipes and cloths, Tork W1 / W2 / W3 / W4
Zásobníky na ubrousky
W1: Tork Performance dispensers for large rolls are ergonomically designed dispensers with safe, reliable tear-off, easy load feature and extra stable construction, which is important in many industrial environments. The floor stand is mobile and easy to move around thanks to its flexible wheels. The wall stand saves space due to its wall mounting.
- Glass fibre reinforced teeth
- Long lasting, durable and sturdy metal construction
- For large paper rolls
W2: The elegant W2 Tork Performance wipe dispenser system dispenses both paper and non woven refills. It provides optimum protection for paper from dirt, damp and theft.
- Tough construction and solid manufacture ensure reliability and durability
- Water resistant seals protect wipes: Improved hygiene and less wastage
- Side-opening hood: Saves refilling time and cost
W3: Tork dispenser for wipe pack rolls is a high capacity dispenser which features easy refilling. Thanks to its large roll capacity, it is ideal for high use areas.
- Optimised one hand dispensing for all needs: Users can take as much as they need or sheet-by-sheet
- Flexible usage: Carton can be put on dispenser shelf or brought to preferred workplace
W4: Tork Performance dispenser for folded wipes is wall mountable and easy to detach. It provides hygienic protection plus 'one at a time' dispensing to reduce consumption and waste.
- Wall bracket for mounting: Convenient installation, facilitates cleaning
- Flexible mounting options with magnets: No drilling needed