Tlačiť
Wipes for delicate tasks, WYPALL* X60
Dodávateľ: KIMBERLY CLARK
6035 8370 6034 8371 8380
115-2166EA 112.31 EUR
115-2166 115-2104 115-2063 115-1279 115-1988
Wipes for delicate tasks, WYPALL* X60
Ubousky/Utěrky Utěrky jednorázové a ručníky Ubrousky pro jemné čištění
Durable uniformly reinforced towels for demanding jobs. Can be used in medium to heavy duty applications.
  • Embossed surface traps and holds dirt and liquids
  • No adhesives or binders; wipes leave nothing behind
  • Tough enough for big jobs, yet soft enough for face and hands
  • Rolls can be used with dispensers 115-0045, 115-1533 or 115-2038
Colour: White or blue