Tlačiť
Process wipes, KIMTECH PREP*
Dodávateľ: KIMBERLY CLARK
7643 7622 7644
115-2222EA 391.1 EUR
115-2222 115-2032 115-1557
Process wipes, KIMTECH PREP*
Ubousky/Utěrky Utěrky čistící pro opakovatelné použití Utěrky pro běžné aplikace
Strong, low-lint wipes for repeat use cleaning with chemicals and solvents. Perfect for printing, pre and post-paint applications, glass and appliance manufacturing, engine maintenance.
  • PP, reusable
  • Great for wiping oils and grease
  • Acid, base, and solvent resistant
  • Roll 115-2222 is compatible with dispensers 115-0045, 115-1533 and 115-2038
Colour: Blue